Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jkamp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106738.1 | NP_001100208.1 | 4 | 743 |
Set Membership: |
G-protein coupled receptor |