Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191774.1 | NP_001178703.1 | 2 | 71 | ||
XM_008765001.1 | XP_008763223.1 | 2 | 157 | |||
XM_008765002.1 | XP_008763224.1 | 2 | 80 |