Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mterfd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199387.1 | NP_955419.1 | 4 | 632 | ||
XM_006241509.2 | XP_006241571.1 | 4 | 658 | |||
GenBank mRNA | BC062080.1 | - | 4 | 632 | 5599618 |