Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam49b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126267.2 | NP_001119739.1 | 6, 7 | 544 | ||
XM_006241632.2 | XP_006241694.1 | 4, 5 | 1656 | |||
XM_006241633.1 | XP_006241695.1 | 5, 6 | 535 | |||
XM_006241634.1 | XP_006241696.1 | 4, 5 | 394 | |||
XM_006241635.2 | XP_006241697.1 | 5, 6 | 515 | |||
XM_008765501.1 | XP_008763723.1 | 6, 7 | 623 | |||
GenBank mRNA | BC166469.1 | - | 6, 7 | 544 | 9036548 | |
FQ212269.1 | - | 5, 6 | 540 |