Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134527.1 | NP_001127999.1 | 6 | 890 | ||
XM_006241552.2 | XP_006241614.1 | 6 | 1718 | |||
GenBank mRNA | BC168688.1 | - | 6 | 890 | 7932577 |