Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241900.2 | XP_006241962.1 | 7 | 805 | ||
XM_006241901.2 | XP_006241963.1 | 7 | 806 | |||
XM_006241902.1 | XP_006241964.1 | 5 | 419 | |||
XM_008765628.1 | XP_008763850.1 | 7 | 2194 | |||
XM_216954.8 | XP_216954.4 | 7 | 804 |