Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srebf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033694.1 | NP_001028866.1 | 11, 12 | 2323 | ||
XM_006242079.2 | XP_006242141.1 | 11, 12 | 2678 | |||
GenBank mRNA | BC101902.1 | - | 11, 12 | 2323 | 7934137 |