Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gxylt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100887.1 | NP_001094357.1 | 4, 5 | 637 | ||
XM_006242192.2 | XP_006242254.1 | 3, 4 | 545 | |||
GenBank mRNA | AY623036.1 | - | 2, 3 | 240 | ||
BC167068.1 | - | 3, 4 | 391 | 8363586 |