Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L3mbtl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033695.1 | NP_001028867.1 | 5 | 574 | ||
XM_008765711.1 | XP_008763933.1 | 5 | 654 | |||
GenBank mRNA | BC101865.1 | - | 5 | 574 | 7938856 |