Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pstpip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106824.2 | NP_001100294.2 | 6 | 529 | ||
XM_006243113.2 | XP_006243175.1 | 6 | 852 | |||
XM_008766223.1 | XP_008764445.1 | 6 | 712 | |||
GenBank mRNA | BC158859.1 | - | 6 | 529 | 7930643 |