Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044253.1 | NP_001037718.1 | 1 | 532 | ||
XM_006243271.2 | XP_006243333.1 | 1 | 1469 | |||
XM_008766324.1 | XP_008764546.1 | 2 | 1188 | |||
XM_008766325.1 | XP_008764547.1 | 2 | 1160 | |||
XM_008766326.1 | XP_008764548.1 | 2 | 1119 | |||
XM_008766327.1 | XP_008764549.1 | 2 | 1091 | |||
GenBank mRNA | DQ489562.1 | - | 1 | 532 |