Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rora | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766408.1 | XP_008764630.1 | 10 | 1839 | ||
XM_008766409.1 | XP_008764631.1 | 10 | 1837 | |||
XM_008766410.1 | XP_008764632.1 | 10 | 1495 |