Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006243594.2 | XP_006243656.1 | 29 | 4354 | ||
XM_006243596.2 | XP_006243658.1 | 29 | 4354 | |||
XM_006243597.2 | XP_006243659.1 | 29 | 4354 | |||
XM_006243598.2 | XP_006243660.1 | 29 | 4354 | |||
XM_006243599.2 | XP_006243661.1 | 28 | 4240 | |||
XM_217233.8 | XP_217233.5 | 29 | 4354 |