Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198755.1 | NP_942050.1 | 655 | |||
XM_006235502.2 | XP_006235564.1 | 6 | 784 | |||
XM_006235503.2 | XP_006235565.1 | 5 | 703 | |||
XM_006235504.2 | XP_006235566.1 | 5 | 703 | |||
XM_006242839.2 | XP_006242901.1 | 6 | 784 | |||
XM_006242840.2 | XP_006242902.1 | 5 | 703 | |||
XM_006242841.2 | XP_006242903.1 | 5 | 703 | |||
GenBank mRNA | AB121446.1 | - | 655 |
Gene Symbol: LOC102549510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766105.1 | XP_008764327.1 | 315 |