Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106896.2 | NP_001100366.2 | 3 | 812 | ||
XM_006244649.1 | XP_006244711.1 | 3 | 812 | |||
XM_008766927.1 | XP_008765149.1 | 4 | 791 |