Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106909.1 | NP_001100379.1 | 5 | 610 | ||
GenBank mRNA | FQ230447.1 | - | 4 | 502 | ||
FQ230685.1 | - | 4 | 502 | |||
FQ232543.1 | - | 4 | 502 |