Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106914.1 | NP_001100384.1 | 8 | 4265 | ||
XM_006244965.2 | XP_006245027.1 | 6 | 2314 | |||
XM_006244966.2 | XP_006245028.1 | 6 | 866 | |||
XM_006244967.2 | XP_006245029.1 | 6 | 1378 | |||
XM_008767084.1 | XP_008765306.1 | 5 | 3060 | |||
XM_008767085.1 | XP_008765307.1 | 5 | 3178 | |||
XM_008767086.1 | XP_008765308.1 | 6 | 871 | |||
XM_008767087.1 | XP_008765309.1 | 6 | 800 | |||
GenBank mRNA | FQ209862.1 | - | 7 | 1411 | ||
FQ228496.1 | - | 7 | 1434 |