Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sumo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009672.1 | NP_001009672.1 | 4 | 239 | ||
GenBank mRNA | BC088322.1 | - | 4 | 239 | 7325996 | |
FQ211636.1 | - | 4 | 276 | |||
FQ214927.1 | - | 4 | 309 | |||
FQ215001.1 | - | 4 | 319 | |||
FQ215643.1 | - | 4 | 284 | |||
FQ224870.1 | - | 4 | 308 | |||
FQ234204.1 | - | 4 | 311 |