Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam46d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106936.1 | NP_001100406.1 | 1 | 471 | ||
XM_006257181.2 | XP_006257243.1 | 4 | 886 | |||
XM_008773335.1 | XP_008771557.1 | 5 | 998 | |||
XM_008773336.1 | XP_008771558.1 | 5 | 764 | |||
XM_008773337.1 | XP_008771559.1 | 5 | 1212 | |||
XM_008773338.1 | XP_008771560.1 | 4 | 884 |