Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs6st2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191726.1 | NP_001178655.1 | 5 | 1305 | ||
XM_006257578.2 | XP_006257640.1 | 3 | 1282 | |||
GenBank mRNA | FQ226114.1 | - | 3 | 934 | ||
FQ230487.1 | - | 3 | 934 |