Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magea8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773233.1 | XP_008771455.1 | 4 | 4685 | ||
XM_228890.3 | XP_228890.1 | 5 | 3153 |