Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767616.1 | XP_008765838.1 | 3, 4 | 403 | ||
GenBank mRNA | BC168182.1 | - | 3, 4 | 335 |