Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab11fip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002727755.3 | XP_002727801.1 | 11 | 2790 | ||
XM_006246071.1 | XP_006246133.1 | 10 | 2754 | |||
XM_220262.6 | XP_220262.5 | 12 | 2925 | |||
GenBank mRNA | BC081997.1 | - | 9 | 1007 | 7130781 |