Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpeb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106992.1 | NP_001100462.1 | 2 | 1159 | ||
XM_006245978.2 | XP_006246040.1 | 2 | 2533 | |||
XM_006245979.2 | XP_006246041.1 | 2 | 2533 | |||
XM_006245980.2 | XP_006246042.1 | 2 | 2533 |