Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286183.1 | NP_001273112.1 | 3 | 826 | ||
XM_003750804.2 | XP_003750852.1 | 3 | 954 | |||
XM_003750806.2 | XP_003750854.1 | 2 | 785 | |||
XM_006246153.2 | XP_006246215.1 | 2 | 3124 | |||
XM_006246158.2 | XP_006246220.1 | 3 | 799 | |||
XM_006246159.2 | XP_006246221.1 | 3 | 779 | |||
XM_006246161.1 | XP_006246223.1 | 2 | 713 | |||
XM_008767653.1 | XP_008765875.1 | 3 | 798 | |||
GenBank mRNA | FQ228105.1 | - | 3 | 812 |