Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irgm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012007.1 | NP_001012007.1 | 3 | 1634 | ||
XM_006246178.2 | XP_006246240.1 | 3 | 1691 | |||
XM_006246179.2 | XP_006246241.1 | 2 | 1550 | |||
GenBank mRNA | BC079106.1 | - | 3 | 1634 | 7125255 |