Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igtp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008765.1 | NP_001008765.1 | 4 | 1278 | ||
XM_006246382.2 | XP_006246444.1 | 4 | 1300 | |||
GenBank mRNA | AY321344.1 | - | 2487 | |||
BC129105.1 | - | 4 | 1278 | |||
BC166569.1 | - | 4 | 1279 | 7319275 |