Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012008.1 | NP_001012008.1 | 6 | 1356 | ||
XM_006246695.2 | XP_006246757.1 | 7 | 1526 | |||
XM_006246696.2 | XP_006246758.1 | 7 | 1523 | |||
XM_006246697.2 | XP_006246759.1 | 6 | 1412 | |||
GenBank mRNA | BC081874.1 | - | 6 | 1356 | 7107522 |