Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107066.1 | NP_001100536.1 | 8 | 843 | ||
GenBank mRNA | BC168671.1 | - | 8 | 919 |