Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigzl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768781.1 | XP_008767003.1 | 1582 | |||
XM_008768782.1 | XP_008767004.1 | 1585 | ||||
XM_008768783.1 | XP_008767005.1 | 1640 | ||||
XM_008768809.1 | XP_008767031.1 | 3 | 698 | |||
XM_008768810.1 | XP_008767032.1 | 3 | 630 | |||
XM_008768811.1 | XP_008767033.1 | 3 | 701 | |||
XM_008768812.1 | XP_008767034.1 | 3 | 664 | |||
GenBank mRNA | DN932759.1 | - | 2 | 419 |