Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233425.2 | XP_006233487.1 | 2 | 324 | ||
XM_008761538.1 | XP_008759760.1 | 3 | 519 |