Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107128.2 | NP_001100598.2 | 6 | 899 | ||
NM_001142945.1 | NP_001136417.1 | 5 | 803 | |||
XM_008768974.1 | XP_008767196.1 | 3 | 1268 | |||
XM_008768975.1 | XP_008767197.1 | 6 | 957 |