Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acad10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249386.2 | XP_006249448.1 | 15 | 2476 | ||
GenBank mRNA | BC168650.1 | - | 15 | 2511 | 7121002 |