Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oasl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009681.1 | NP_001009681.1 | 6 | 1115 | ||
GenBank mRNA | AY227756.1 | - | 6 | 1115 | ||
FQ231843.1 | - | 1 | 36 |