Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oasl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009682.1 | NP_001009682.1 | 4 | 849 | ||
XM_008769327.1 | XP_008767549.1 | 4 | 1023 | |||
GenBank mRNA | AY237116.1 | - | 4 | 849 |