Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ep400 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107149.1 | NP_001100619.1 | 27 | 5348 | ||
XM_006249549.2 | XP_006249611.1 | 29 | 5702 | |||
XM_006249550.2 | XP_006249612.1 | 29 | 5867 | |||
XM_006249552.2 | XP_006249614.1 | 29 | 5683 | |||
XM_006249553.2 | XP_006249615.1 | 29 | 5703 | |||
XM_006249554.2 | XP_006249616.1 | 29 | 5702 | |||
XM_006249556.2 | XP_006249618.1 | 28 | 5594 | |||
XM_006249557.2 | XP_006249619.1 | 28 | 5591 | |||
XM_006249558.2 | XP_006249620.1 | 28 | 5459 | |||
XM_006249559.2 | XP_006249621.1 | 26 | 5239 | |||
XM_006249560.2 | XP_006249622.1 | 28 | 5590 | |||
XM_008769332.1 | XP_008767554.1 | 30 | 5804 |