Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtbdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107165.1 | NP_001100635.1 | 4 | 838 | ||
XM_008772381.1 | XP_008770603.1 | 5 | 962 | |||
XM_008772382.1 | XP_008770604.1 | 5 | 977 | |||
XM_008772383.1 | XP_008770605.1 | 5 | 959 | |||
XM_008772384.1 | XP_008770606.1 | 5 | 936 | |||
XM_008772385.1 | XP_008770607.1 | 6 | 898 | |||
XM_008772386.1 | XP_008770608.1 | 6 | 773 | |||
XM_008772387.1 | XP_008770609.1 | 5 | 791 | |||
XM_008772388.1 | XP_008770610.1 | 5 | 908 | |||
XM_008772389.1 | XP_008770611.1 | 5 | 885 | |||
XM_008772390.1 | XP_008770612.1 | 5 | 904 | |||
XM_008772391.1 | XP_008770613.1 | 5 | 889 | |||
XM_008772392.1 | XP_008770614.1 | 5 | 603 | |||
XM_008772393.1 | XP_008770615.1 | 5 | 912 | |||
XM_008772394.1 | XP_008770616.1 | 5 | 692 | |||
XM_008772395.1 | XP_008770617.1 | 5 | 685 | |||
XM_008772396.1 | XP_008770618.1 | 5 | 765 | |||
XM_008772397.1 | XP_008770619.1 | 5 | 718 | |||
XM_008772398.1 | XP_008770620.1 | 5 | 666 |