Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hook2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100562.1 | NP_001094032.1 | 9, 10 | 841 | ||
XM_006255255.2 | XP_006255317.1 | 9, 10 | 972 | |||
XM_006255257.2 | XP_006255319.1 | 9, 10 | 971 | |||
XM_006255258.2 | XP_006255320.1 | 9, 10 | 973 | |||
XM_008772399.1 | XP_008770621.1 | 9, 10 | 974 | |||
GenBank mRNA | BC099235.1 | - | 4, 5 | 379 |