Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107167.1 | NP_001100637.1 | 3 | 405 | ||
XM_006249632.2 | XP_006249694.2 | 2 | 555 | |||
XM_006249633.2 | XP_006249695.2 | 2 | 257 |