Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3hdm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134867.1 | NP_001128339.1 | 10 | 1188 | ||
XM_006249690.2 | XP_006249752.1 | 10 | 1548 | |||
XM_006249694.2 | XP_006249756.1 | 8 | 1053 | |||
XM_006249698.1 | XP_006249760.1 | 3 | 271 | |||
XM_008769459.1 | XP_008767681.1 | 10 | 1185 | |||
XM_008769460.1 | XP_008767682.1 | 10 | 1185 | |||
XM_008769461.1 | XP_008767683.1 | 10 | 1185 | |||
XM_008769462.1 | XP_008767684.1 | 10 | 1185 | |||
XM_008769463.1 | XP_008767685.1 | 8 | 1052 | |||
GenBank mRNA | BC087715.1 | - | 9 | 866 |