Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pogk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107194.2 | NP_001100664.2 | 5 | 990 | ||
XM_006250161.2 | XP_006250223.1 | 6 | 1364 | |||
XM_006250162.2 | XP_006250224.1 | 6 | 1229 |