Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shroom3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100889.1 | NP_001094359.1 | 7 | 3994 | ||
XM_006250723.2 | XP_006250785.1 | 7 | 5005 | |||
XM_006250725.2 | XP_006250787.1 | 5 | 6778 |