Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004098.1 | NP_001004098.1 | 8 | 929 | ||
GenBank mRNA | AY693787.1 | - | 3 | 353 | ||
AY693788.1 | - | 8 | 929 |