Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107217.1 | NP_001100687.1 | 2 | 226 | ||
GenBank mRNA | FQ211206.1 | - | 2 | 188 | ||
FQ218099.1 | - | 2 | 193 | |||
FQ221610.1 | - | 2 | 190 |