Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017374.2 | NP_001017374.2 | 6, 7 | 792 | ||
GenBank mRNA | BC099117.1 | - | 6, 7 | 679 |
Gene Symbol: LOC501116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765067.1 | XP_008763289.1 | 2 | 614 |