Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271341.1 | NP_001258270.1 | 1 | 93 | ||
XM_006252893.2 | XP_006252955.1 | 1 | 578 | |||
XM_006252894.2 | XP_006252956.1 | 1 | 578 | |||
XM_008771104.1 | XP_008769326.1 | 1 | 578 | |||
GenBank mRNA | EU861029.1 | - | 1 | 105 |