Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012050.1 | NP_001012050.1 | 6 | 1173 | ||
XM_006253091.2 | XP_006253153.1 | 6 | 976 | |||
GenBank mRNA | BC088452.1 | - | 6 | 1173 | 7126921 | |
FQ232328.1 | - | 6 | 1230 |