Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134863.1 | NP_001128335.1 | 3 | 2997 | ||
XM_006253151.2 | XP_006253213.2 | 3 | 2280 | |||
XM_006253152.2 | XP_006253214.1 | 3 | 1708 | |||
XM_006253153.2 | XP_006253215.1 | 3 | 2088 | |||
XM_008771258.1 | XP_008769480.1 | 3 | 1848 |