Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008340.1 | NP_001008341.1 | 1 | 266 | ||
GenBank mRNA | BC086347.1 | - | 1 | 266 | 7302510 | |
JF423321.1 | - | 1 | 266 |