Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191692.1 | NP_001178621.1 | 27 | 2313 | ||
XM_008771640.1 | XP_008769862.1 | 19 | 1912 | |||
XM_008771641.1 | XP_008769863.1 | 19 | 1912 | |||
XM_008771642.1 | XP_008769864.1 | 18 | 1900 | |||
XM_008771643.1 | XP_008769865.1 | 19 | 1912 | |||
XM_008771644.1 | XP_008769866.1 | 18 | 1900 | |||
XM_008771645.1 | XP_008769867.1 | 19 | 1912 | |||
XM_008771646.1 | XP_008769868.1 | 19 | 1912 | |||
XM_008771647.1 | XP_008769869.1 | 18 | 1899 | |||
XM_008771648.1 | XP_008769870.1 | 19 | 1912 | |||
XM_008771649.1 | XP_008769871.1 | 19 | 1912 | |||
XM_008771650.1 | XP_008769872.1 | 19 | 1912 | |||
XM_008771651.1 | XP_008769873.1 | 19 | 1912 | |||
XM_008771652.1 | XP_008769874.1 | 19 | 1912 |